missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl sulfide, 98%, Thermo Scientific Chemicals
282.10 SEK - 716.52 SEK
Chemical Identifiers
CAS | 592-88-1 |
---|---|
Molecular Formula | C6H10S |
Molecular Weight (g/mol) | 114.21 |
MDL Number | MFCD00008658 |
InChI Key | UBJVUCKUDDKUJF-UHFFFAOYSA-N |
Synonym | diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide |
PubChem CID | 11617 |
ChEBI | CHEBI:4489 |
IUPAC Name | 3-prop-2-enylsulfanylprop-1-ene |
SMILES | C=CCSCC=C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-88-1 | |
114.21 | |
UBJVUCKUDDKUJF-UHFFFAOYSA-N | |
11617 | |
3-prop-2-enylsulfanylprop-1-ene |
C6H10S | |
MFCD00008658 | |
diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide | |
CHEBI:4489 | |
C=CCSCC=C |
Specifications
592-88-1 | |
100.0 | |
Colorless | |
138.0°C | |
Authentic | |
C6H10S | |
1.4879 to 1.4899 | |
01, 440 | |
0.887 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether and,carbon tetrachloride | |
C=CCSCC=C | |
114.21 | |
CHEBI:4489 | |
98% | |
Allyl sulfide |
96.0 | |
-83.0°C | |
0.8870g/mL | |
46°C | |
98% | |
(H2C=CHCH2)2S | |
MFCD00008658 | |
15, 288 | |
diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide | |
UBJVUCKUDDKUJF-UHFFFAOYSA-N | |
3-prop-2-enylsulfanylprop-1-ene | |
11617 | |
114.2 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-775-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Allyl sulfide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only