missing translation for 'onlineSavingsMsg'
Learn More

alpha-(4-Morpholinyl)phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Product Code. 11308775
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15190-10-0
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.26
MDL Number MFCD00014619
InChI Key MWZPYQLYZXTCLZ-UHFFFAOYNA-N
Synonym 2-morpholino-2-phenylacetonitrile, morpholin-4-yl phenyl acetonitrile, 2-morpholin-4-yl-2-phenylacetonitrile, alpha-4-morpholino phenylacetonitrile, morpholino phenyl acetonitrile, 4-morpholineacetonitrile, .alpha.-phenyl, morpholin-4-yl-phenyl-acetonitrile, 2-morpholin-4-yl-2-phenylethanenitrile, alpha-4-morpholinyl phenylacetonitrile, maybridge1_000080
PubChem CID 85812
IUPAC Name 2-morpholin-4-yl-2-phenylacetonitrile
SMILES N#CC(N1CCOCC1)C1=CC=CC=C1

Specifications

Melting Point 67°C to 71°C
Quantity 25 g
UN Number UN3439
Beilstein 181704
Formula Weight 202.26
Percent Purity ≥98%
Chemical Name or Material alpha-(4-Morpholinyl)phenylacetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'