missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Mouse anti-Human, Clone: ACTN2/3291, Novus Biologicals™
Mouse Monoclonal Antibody
2450.00 SEK
Specifications
Antigen | Alpha Actinin 2 |
---|---|
Clone | ACTN2/3291 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Alpha Actinin 2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein(exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
ACTN2/3291 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
88 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title