missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Fodrin Mouse anti-Human, Clone: SPTAN1/3505, Novus Biologicals™
Mouse Monoclonal Antibody
2470.00 SEK - 4910.00 SEK
Specifications
Antigen | Alpha Fodrin |
---|---|
Clone | SPTAN1/3505 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18332512
|
Bio-Techne
NBP3-13781-20UG |
20 μg |
2470.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18321517
|
Bio-Techne
NBP3-13781-100UG |
100 μg |
4910.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Alpha Fodrin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Neuroscience | |
10 mM PBS with 0.05% BSA | |
Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
Recombinant fragment of human Alpha Fodrin protein (around aa2351-2475) (exact sequence is proprietary)) (Uniprot: Q13813) | |
Primary | |
Store at 4°C. |
SPTAN1/3505 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q13813 | |
6709 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title