missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulphate, Scharlab
Click to view available options
IUPAC Name:
dialuminium(3+) nonahydrate trisulfate
Quantity:
1 kg
Specifications
Specifications
CAS | 7784-31-8 |
Quantity | 1 kg |
Molecular Formula | Al2H18O21S3 |
InChI Key | KFCWRPFNNIEZRY-UHFFFAOYSA-H |
SMILES | O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
IUPAC Name | dialuminium(3+) nonahydrate trisulfate |
Molecular Weight (g/mol) | 504.27 |
Grade | Pharmpur |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction