missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, ACS Reagent, 99%, Honeywell™ Fluka™

Product Code. 15633170
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Boiling Point 2460°C
Melting Point 660.37°C
CAS 7429-90-5
Quantity 100 g
Molecular Formula Al
MDL Number MFCD00134029
UN Number UN3077
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES [Al]
IUPAC Name aluminium
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
ChEBI CHEBI:33629
Formula Weight 26.98g/mol
Percent Purity 99%
Grade ACS Reagent
Ignition Point 1400°F
Packaging Plastic bottle
Density 2.7g/mL at 25°C
Chemical Name or Material Aluminum
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'