missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Fluoride Trihydrate, 97%, Honeywell Fluka™

Product Code. 15642330
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00149141
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]

Specifications

Color White
Quantity 500 g
Linear Formula AlF3·3H2O
UN Number NONH for all modes of transport
Formula Weight 138.02g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 2.88g/mL at 25°C
Chemical Name or Material Aluminum Fluoride Trihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'