missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Aluminum Reagent Set, Aluminon

Reagent set for determination of aluminum by the AluVer™3 Aluminon method.

Brand:  Hach Pacific Pty Ltd 2242000

Product Code. 16805673

  • 4795.00 SEK / 100 tests
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reagent set for determination of aluminum by the AluVer™3 Aluminon method.Hach Method 8012. Range: 0.008 - 0.800 mg/L Al.Set contains 100 each: AluVer™3 Aluminon Reagent Powder Pillows, Ascorbic Acid Powder Pillows and Bleaching 3 Reagent Powder Pillows.100 tests.

    Specifications

  • Description : Aluminum Reagent Set, Powder Pillows, Aluminon
  • Instrument: All
  • Method: 8012
  • Method Name: Aluminon
  • Number of tests: 100
  • Parameter: Aluminum
  • Platform: Powder Pillows
  • Range: 0.008 - 0.800 mg/L Al
  • Special Remarks: Set contains: AluVer 3 aluminum Powder Pillows (1429099), Ascorbic Acid Powder Pillows(1457799), Bleaching 3 Powder Pillows (1429449)
  • Storage conditions: 10 °C to 30 °C
Specifications

Specifications

100 Tests
0.008 to 0.800 mg/L Al
Powder Pillows
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers