Learn More
Eppendorf™ Aluminum slide-out platform for I26 and I26R shakers
Eppendorf shaker platforms are available in a wide variety of materials and sizes, perfect for virtually every requirement. Universal platforms can be adapted for thousands of combinations of clamps and other accessories. Dedicated platforms come complete with a single clamp size pre-installed.
Brand: Eppendorf™ M1324-9907
This item is not returnable.
View return policy
Description
Aluminum slide-out platform for I26 and I26R shakers, 76 × 46 cm, not interchangeable with other 76 x 46 cm platforms, 500 mL Erlenmeyer flask dedicated platform- Flask Capacity: 24Specifications
76 x 46 cm | |
Not interchangeable with others | |
500 mL Erlenmeyer Flask Dedicated Platform |