missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate

Amber glass to protect light-sensitive contents. Delivered with individual USP conformity certificate.

Brand:  BRAND™ 957488

780.21 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15871208

  • 885.00 SEK / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Delivered with individual USP conformity certificate
  • Class A, DE-M marking
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
200 mL
1
NS 14/23
A
15.5 mm
Borosilicate Glass (3.3)
Volumetric flask
±0.10 mL
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate > Capacity: 200mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.