Learn More
Invitrogen™ Retic Lysate IVT™ Kit
Efficiently translates high molecular weight proteins and both capped and uncapped mRNAs
Brand:  Invitrogen™ AM1200M
 View more versions of this product
Product Code. 10005314
Includes: Retic Lysate should be stored at -80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below -70°C. Nuclease-free Water can be stored at any temperature.
Description
Description
The Retic Lysate IVT™ Kit efficiently translates in vitro-synthesized transcripts, poly(A) RNA, and total RNA, including difficult-to-translate RNAs. The optimized lysate results in high protein yield, biological activity, and consistent performance. A report in Nucleic Acids Research (1) found that some commercial reticulocyte lysates had poor fidelity for initiation at the appropriate AUG initiator codon. The Retic Lysate IVT Kit was tested using Kozak's constructs and it demonstrated high fidelity for initiating at the proper AUG.
  • Efficiently translates a wide variety of high molecular weight proteins and both capped and uncapped mRNAs
  • Accommodates large volume of template mRNA
How it works
The lysate has been optimized for translation by the addition of an ATP-regenerating system, hemin, and calf liver tRNA. Included in the kit is an optimized Translation Mix (-Met or -Leu) for the translation of both capped and uncapped messages along with High and Low Salt buffers. These buffers can be used to maximize protein yield and activity from difficult-to-translate sequences.
1. Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res 18:2828.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Retic Lysate should be stored at –80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below –70°C. Nuclease-Free water can be stored at any temperature.
Water Soluble Protein
Cell-Free
60 Reactions
Retic Lysate with Manual
Kit
Cell-Free Expression
In Vitro Transcription
Ambion™
60 Reactions
Product Title
Invitrogen™ Retic Lysate IVT™ Kit > 60 reactions; With manual