Learn More
Ammonium dihydrogen arsenate, 98%, Thermo Scientific Chemicals
3290.00 SEK - 13845.00 SEK
Chemical Identifiers
CAS | 13462-93-6 |
---|---|
Molecular Formula | AsH6NO4 |
Molecular Weight (g/mol) | 158.973 |
MDL Number | MFCD00064864 |
InChI Key | VTWJXWNBCACAAK-UHFFFAOYSA-N |
Synonym | ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1 |
PubChem CID | 166814 |
IUPAC Name | azanium;dihydrogen arsorate |
SMILES | [NH4+].O[As](=O)(O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383897
|
Thermo Scientific Alfa Aesar
031493.18 |
50 g |
3290.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373897
|
Thermo Scientific Alfa Aesar
031493.30 |
250 g |
13845.00 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13462-93-6 | |
158.973 | |
VTWJXWNBCACAAK-UHFFFAOYSA-N | |
166814 | |
[NH4+].O[As](=O)(O)[O-] |
AsH6NO4 | |
MFCD00064864 | |
ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1 | |
azanium;dihydrogen arsorate |
Specifications
300°C (decomposition) | |
Crystalline | |
MFCD00064864 | |
ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1 | |
VTWJXWNBCACAAK-UHFFFAOYSA-N | |
azanium;dihydrogen arsorate | |
166814 | |
98% | |
2.311 g/mL |
13462-93-6 | |
AsH6NO4 | |
UN1557 | |
Soluble in water. (337.4mg/ml at 0°C). | |
[NH4+].O[As](=O)(O)[O-] | |
158.973 | |
158.98 | |
1.577 | |
Ammonium dihydrogen arsenate |
Safety and Handling
GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 236-667-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.