missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10191890

Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10191890
1 kg
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10191890

Brand: Fisher Chemical A/5760/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H24MO7N6O24.4H2O, CAS Number-12054-85-2, 500g, White, 1235.86g/mol, 190 deg.C, 167059, Plastic powder jar, 5.0 to 5.5, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-85-2
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
MDL Number 167059
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Melting Point 190°C
Color White
pH 5.0 to 5.5
Physical Form Solid
Quantity 1 kg
Formula Weight 1235.86g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Ammonium molybdate(VI) tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.