missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, MP Biomedicals™

Product Code. 11490492
Click to view available options
Quantity:
500 g
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the precipitation or fractionation of proteins and purification of antibodies. Ammonium Sulfate, MP Biomedicals is an inorganic salt useful in crystallographic analysis of nucleic acids and proteins. Use in long PCR buffer, PCR lysis solution and

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble freely in water; insoluble in alcohol, acetone.
Formula Weight 132.1
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.