missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™ Product Code.: 15645510

Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™

Product Code. 15645510
250 g, Poly Bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15645510

Brand: Honeywell Research Chemicals 431354250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium saltShow MoreShow Less
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 140°C to 149°C
Color White
pH 4.5 to 6
Physical Form Solid
Quantity 250 g
Linear Formula NH4SCN
Beilstein 3595135
Solubility Information In water 1630g/L at 20°C
Formula Weight 76.12g/mol
Percent Purity 99.99% (trace metals basis)
Packaging Poly Bottle
Appearance Crystals
Density 1.3g/cm3
Chemical Name or Material Ammonium thiocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.