Learn More
Hettich Lab Technology™ Angle Rotor for ROTINA 420/420 R, 4-place
Angle Rotor well equipped for all centrifuging applications. ROTINA 420 Centrifuges are ideal for applications in clinical chemistry, in biotechnology and in life science research facilities.
Brand: Hettich Lab Technology™ 4795
This item is not returnable.
View return policy
Description
- Angle Rotor
- 25°, 4-place
- For use with ROTINA 420/420 R Centrifuge
Specifications
Angle Rotor | |
9,500 rpm | |
100 mm | |
61 mm | |
25° |
12,007 x g | |
119 mm | |
122 mm | |
ROTINA 420/420 R | |
4 |