Learn More
ANGPTL4 Mouse anti-Human, Unconjugated, Clone: 1D2H6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67577-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16870286
  • 4721.67 SEK / 150µL
In Stock
to see stock.
Description
Description
The ANGPTL4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANGPTL4. This antibody recognizes human, mouse antigen. The ANGPTL4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ANGPTL4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, ELISA
1D2H6
Unconjugated
150 μL
51129
IgG2b