missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A10 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518004
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.Specifications
Annexin A10 | |
Polyclonal | |
Unconjugated | |
ANXA10 | |
annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
11199 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UJ72 | |
ANXA10 | |
Synthetic peptide sequence (FCGDYVQGTIFPAPN) corresponding to the N-terminus amino acids of ANXA10 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |