Learn More
Bcl-2 Mouse anti-Human, PE, Clone: BCL-2/100, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340576
Description
Anti-bcl-2, clone 100, is derived from the fusion of Sp2/0-Ag14 cells with spleen cells from BALB/c mice immunized with the bcl-2α peptide (amino acids 41-54).
Anti–bcl-2 recognizes a 26-kilodalton (kd) protein encoded by the 230 kb bcl-2 protooncogene. The protein is a regulator of the apoptotic process, and elevated levels can provide resistance to cell death. The bcl-2 protein is expressed in the interior of cells, frequently localized to the mitochondrial membrane.
Specifications
Bcl-2 | |
Monoclonal | |
1μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human Bcl-2 synthetic peptide aa. 41-54 | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry, Immunocytochemistry | |
BCL-2/100 | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.