Learn More
anti-BUB1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000699-AP22.1Set
Product Code. 16191794
  • 10645.00 SEK / 1 set
Explore more special offers
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-BUB1 (100μg) , Detection antibody: mouse purified polyclonal anti-BUB1 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy
Specifications
BUB1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BUB1.
BUB1A/BUB1L/hBUB1
Mouse, Rabbit
RUO
699
Human
Liquid
ELISA
Unconjugated
BUB1
BUB1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-BUB1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-BUB1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.