missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sensing Receptor Monoclonal Antibody (5C10, ADD), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1934
This item is not returnable.
View return policy
Description
MA1-934 detects human, bovine, mouse, and rat calcium sensing receptor. MA1-934 has been successfully used in Western blot, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ∽ 150 and 130 kDa proteins from human HEK-293 cells.This antibody has also been used in immunohistochemical on frozen bovine parathyroid gland and rat thyroid gland. The MA1-934 immunogen is a synthetic peptide corresponding to residues (214) A D D D Y G R P G I E K F RE E A E E R D I (235) of human calcium sensing receptor.
Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.Specifications
Calcium Sensing Receptor | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P35384, P41180, P48442, Q9QY96 | |
CASR | |
Synthetic peptide corresponding to residues (214) A D D D Y G R P G I E K F RE E A E E R D I (235) of human calcium sensing receptor. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
5C10, ADD, ADD | |
Unconjugated | |
CASR | |
BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1 | |
Mouse | |
Protein A | |
RUO | |
12374, 24247, 281038, 846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |