missing translation for 'onlineSavingsMsg'
Learn More

CD55 Monoclonal Antibody (583905), Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen MA524264

Product Code. 15573952

  • 4180.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

In direct ELISAs, no cross-reactivity with recombinant human CD55 or recombinant mouse CD97 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

CD55 Monoclonal antibody specifically detects CD55 in Mouse samples. It is validated for Flow Cytometry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD55
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q61475
CD55
Mouse myeloma cell line NS0-derived recombinant mouse CD55/DAF Met1-Pro359
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry, Western Blot
583905
Unconjugated
CD55
CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC
Rat
Protein A/G
RUO
13136
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers