missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FABP1 Ab-1 Matched Antibody Pair, Abnova™
Brand: Abnova H00002168-AP12.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FABP1 (100μg); Detection antibody: mouse monoclonal anti-FABP1, IgG2b κ (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FABP1
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2b κ
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
FABP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FABP1. | |
FABPL/L-FABP | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FABP1 (100 μg) 2. Detection antibody: mouse monoclonal anti-FABP1, IgG2b Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
FABP1 | |
FABP1 | |
Affinity Purified | |
RUO | |
2168 | |
Human | |
IgG, IgG2b κ |