Learn More
FABP2, Mouse, Clone: 2F7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FABP2.
Brand: Abnova H00002169-M11.100ug
Description
The intracellular fatty acid-binding proteins (FABPs) belong to a multigene family with nearly twenty identified members. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Intestinal fatty acid-binding protein 2 gene contains four exons and is an abundant cytosolic protein in small intestine epithelial cells. This gene has a polymorphism at codon 54 that identified an alanine-encoding allele and a threonine-encoding allele. Thr-54 protein is associated with increased fat oxidation and insulin resistance. [provided by RefSeq
Sequence: MAFDSTWKVDRSENYDKFMEKMGVNIVKRKLAAHDNLKLTITQEGNKFTVKESSAFRNIEVVFELGVTFNYNLADGTELRGTWSLEGNKLSpecifications
FABP2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000134 | |
FABP2 | |
FABP2 (NP_000125.1, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
2F7 | |
Mouse monoclonal antibody raised against a partial recombinant FABP2. | |
FABP2 | |
FABPI/I-FABP/MGC133132 | |
Mouse | |
Affinity chromatography | |
RUO | |
2169 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |