missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flotillin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521296
3556.00 SEK valid until 2025-03-16
Use promo code "24070" to get your promotional price.
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
Specifications
Flotillin 2 | |
Polyclonal | |
Unconjugated | |
FLOT2 | |
AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14252, 2319, 83764 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q14254, Q60634, Q9Z2S9 | |
FLOT2 | |
Synthetic peptide corresponding to a region within amino acids 365 and 428 of Flotillin 2 (Uniprot ID#Q14254) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Flotillin 2 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction