Learn More
GEF-H1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA538917
Product Code. 15515831
  • 4336.30 SEK / 100µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate rho-dependent signals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GEF-H1
Polyclonal
Unconjugated
ARHGEF2
GEF, GEF-H1, GEFH1, LFP40, P40, Rho/Rac guanine nucleotide exchange factor 2, guanine nucleotide exchange factor H1, microtubule-regulated Rho-GEF, proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2
Rabbit
Antigen affinity chromatography
RUO
16800, 9181
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q92974, Q60875
ARHGEF2
A synthetic non-phosphopeptide derived from human Rho/Rac Guanine Nucleotide Exchange Factor 2 around the phosphorylation site of Ser885 (R-R-SP-L-P)
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GEF-H1 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated