Learn More
GPBB Mouse anti-Human, Clone: 17B6, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121527
Additional Details : Weight : 0.01000kg
Description
MA1-21527 detects GPBB in Human samples. MA1-21527 has been successfully used in ELISA and Western Blot procedures. MA1-21527 immunogen corresponds to Full length protein (Human)..
GPBB - sensitive marker for the AMI diagnosis within 4 hours after the onset of chest pain. It has been also shown that GPBB increased in a considerable proportion of AMI patients within 2-3 hours from chest pain onset. GPBB increased early in patients with unstable angina. GPBB can be also a sensitive marker for the detection of peri-operative myocardial ischaemia and infraction in patients undergoing coronary artery bypass grafting.Specifications
GPBB | |
Monoclonal | |
Unconjugated | |
P11216 | |
PYGB | |
Full length protein (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
17B6 | |
PYGB | |
MGC9213 | |
Mouse | |
Protein A | |
RUO | |
5834 | |
4° C, do not freeze | |
Liquid |