missing translation for 'onlineSavingsMsg'
Learn More

HSP90 alpha Monoclonal Antibody (3B6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA3010

Product Code. 11505732

  • 4530.00 SEK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MA3-010 detects heat shock protein 90 kDa (HSP90) from human, mouse, rat, and bovine tissues. This antibody does not detect chicken HSP90. MA3-010 has been successfully used in Western blot, immunofluorescence, and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects a 90 kDa protein representing HSP90 from MCF7 cell extract. MA3-011 is more sensitive in Western blot procedures and should be used when maximum sensitivity is desired. MA3-010 does not immunoprecipitate HSP90 in any form (free or complexed). The MA3-010 antigen is purified mouse HSP90.

Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSP90 alpha
Monoclonal
Conc. Not Determined
ascites diluted in PBS with 0.05% sodium azide
P07900, P07901, P82995, Q76LV2
Hsp90aa1
Purified mouse HSP90.
RUO
15519, 281832, 299331, 3320
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
3B6
Unconjugated
Hsp90aa1
86kDa; 89kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; akineto; AL024080; AL024147; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; fb17b01; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90; heat shock protein 90 alpha family class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1, tandem duplicate 1; heat shock protein 90-alpha 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-alpha 1; heat shock protein HSP90 alpha; heat shock protein hsp90a
Mouse
100 μL
Primary
Bovine, Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers