missing translation for 'onlineSavingsMsg'
Learn More

Lamin B1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA519468

Product Code. 11387433

  • 4550.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with chicken and zebrafish based on sequence homology.

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Defects in the LMNB1 gene can cause Leukodystrophy, demyelinating, autosomal dominant, adult-onset (ADLD).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lamin B1
Polyclonal
Unconjugated
Lmnb1
0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg
Rabbit
Antigen affinity chromatography
RUO
116685, 16906, 4001
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.02% sodium azide; pH 7.4
P14733, P20700, P70615
Lmnb1
Synthetic peptide conjugated to KLH derived from within residues 400 - 500 of Mouse Lamin B1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers