missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIB2 Monoclonal Antibody (739501), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524352
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
MIB2 Monoclonal antibody specifically detects MIB2 in Human samples. It is validated for Immunocytochemistry.Specifications
MIB2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96AX9 | |
MIB2 | |
E. coli-derived recombinant human Mind Bomb 2/MIB2 Ala418-Lys520 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry | |
739501 | |
Unconjugated | |
MIB2 | |
2210008I11Rik; Dyslike; dystrophin-like protein; E3 ubiquitin-protein ligase MIB2; EC 6.3.2.13; FLJ20648; FLJ41406; Mib2; Mind bomb homolog 2; mind bomb-2; mindbomb E3 ubiquitin protein ligase 2; mindbomb homolog 2; mindbomb homolog 2 (Drosophila); novel zinc finger protein; Novelzin; putative NF-kappa-B-activating protein 002N; RBSC-skeletrophin; RBSC-skeletrophin/dystrophin-like polypeptide; RING-type E3 ubiquitin transferase MIB2; skd; Skeletrophin; zinc finger ZZ type with ankyrin repeat domain protein 1; zinc finger, ZZ type with ankyrin repeat domain 1; Zzank1; ZZZ5 | |
Mouse | |
Protein A/G | |
RUO | |
142678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction