Learn More
MMP10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA124316
Product Code. 11530963
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
PA1-24316 detects MMP-10 in Human samples. PA1-24316 has been successfully used in Western Blot procedures. PA1-24316 immunogen corresponds to a synthetic peptide corresponding to a region of human MMP10. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
MMP-10 can degrade fibronectin, gelatins of type I, III, IV, and V; weakly collagens III, IV, and V. It activates procollagenase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP10
Polyclonal
Unconjugated
P09238
MMP10
Synthetic peptide region of human MMP10.
100 μg
Primary
Human
Antibody
IgG
Western Blot
1 mg/mL
MMP10
SL-2, STMY2
Rabbit
Protein A/G
RUO
4319
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MMP10 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated