missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH11 Monoclonal Antibody (SM-M10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511971
This item is not returnable.
View return policy
Description
MA5-11971 targets Myosin Smooth Muscle Heavy Chain in IF and IHC (P) applications and shows reactivity with Bovine and Human samples. The MA5-11971 immunogen is crude human uterus extract.
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.Specifications
MYH11 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O08638, P35749 | |
MYH11 | |
Crude human uterus extract | |
500 μL | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
SM-M10 | |
Unconjugated | |
MYH11 | |
AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain | |
Mouse | |
Protein G | |
RUO | |
17880, 4629, 530050 | |
4° C | |
Liquid |