missing translation for 'onlineSavingsMsg'
Learn More

OLFM1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA528842

3003.48 SEK valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 13492957

  • 4505.00 SEK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HepG2, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OLFM1
Polyclonal
Unconjugated
OLFM1
1B426B; AMY; AW742568; D2Sut1e; neuroblastoma protein; neuronal olfactomedin-related ER localized protein; Noe1; Noel; Noel1; Noelin; Noelin 1; Noelin 2; NOELIN1; OlfA; olfactomedin 1; olfactomedin related ER localized protein; olfactomedin-1; OLFM1; Pancortin; pancortin 1; Pancortin 1-4; Pancortin3
Rabbit
Antigen affinity chromatography
RUO
10439, 56177
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O88998, Q99784
OLFM1
Synthetic peptide corresponding to a region within amino acids 421 and 485 of Human Noelin
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
OLFM1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.