Learn More
PTEN, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00005728-PW2.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-PTEN (300μg), Antibody pair for WB: mouse purified polyclonal anti-PTEN (50μg)
Specifications
PTEN | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
10q23del/BZS/MGC11227/MHAM/MMAC1/PTEN1/TEP1 | |
Mouse | |
RUO | |
5728 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PTEN | |
PTEN | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PTEN (300 ug) 2. Antibody pair for WB: mouse purified polyclonal anti-PTEN (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTEN, Human, IP-WB Antibody Pair (PW2), Abnova™ > 1 Set