missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM6 Monoclonal Antibody (3E9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520951
This item is not returnable.
View return policy
Description
Peptide Sequence: DSPERKRIKY SRETDSDRKL VDKEDIDTSS KGGCVQQATG WRKGTGLGYG HPGLASSEEA EGRMRGPSVG ASGRTSKRQS NETYRDAVRR VMFARYKELD
RBM6 Monoclonal antibody specifically detects RBM6 in Human samples. It is validated for ELISA, Immunohistochemistry (PFA fixed), Western Blot.Specifications
RBM6 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P78332 | |
RBM6 | |
RBM6 (NP_005768.1, 1024 a.a. ∽ 1123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
3E9 | |
Unconjugated | |
RBM6 | |
3G2; 4930506F14Rik; DEF3; DEF-3; ETK2; g16; HEK2; HLC-11; Lung cancer antigen NY-LU-12; lung cancer protooncogene 11; mKIAA4015; NY-LU-12; Protein G16; Rbm6; RGD1560367; RNA binding motif protein 6; RNA-binding motif protein 6; RNA-binding protein 6; RNA-binding protein DEF-3; TYRO6 | |
Mouse | |
Affinity chromatography | |
RUO | |
10180 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction