missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519319
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes an enzyme belonging to the short-chain dehydrogenases/reductases family. This retinol dehydrogenase functions to catalyze the final step in the biosynthesis of 11-cis retinaldehyde, which is the universal chromophore of visual pigments. Mutations in this gene cause autosomal recessive fundus albipunctatus, a rare form of night blindness that is characterized by a delay in the regeneration of cone and rod photopigments. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring upstream BLOC1S1 gene.
Specifications
RDH5 | |
Polyclonal | |
Unconjugated | |
RDH5 | |
11-cis RDH; 11-cis retinol dehydrogenase; 11-cis RoDH; 9-cis; 9-cis retinol dehydrogenase; 9-cis-retinol dehydrogenase; 9-cis-retinol specific dehydrogenase; 9cRDH; AI987873; Cis-retinol dehydrogenase; cRDH; FLJ39337; FLJ97089; HSD17B9; rdh; RDH1; Rdh4; rdh5; retinol dehydrogenase 1; retinol dehydrogenase 5; retinol dehydrogenase 5 (11-cis and 9-cis); retinol dehydrogenase 5 (11-cis/9-cis); retinol dehydrogenase type 5; SDR9C5; Short chain dehydrogenase/reductase family 9C member 5; short chain dehydrogenase/reductase family 9C, member 5 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q92781 | |
RDH5 | |
Synthetic peptide sequence (RDRQSLPASNAFV) corresponding to the internal amino acids of RDH5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RDH5 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction