Learn More
Brady™ Anti-skid Laminated Polyester Anti-skid Tape
Brand:  Brady™ 078092
Product Code. 11741645
Specifications
Roll-Mounted Anti-Skid Tape - Yellow
Traction on steps, slick floors, and egress pathway marking
1 Roll
0.162 mm
Permanent Acrylic
Polyester
18 m x 100 mm
18 m
-40°C to +80°C
100 mm
Yellow
Anti-Skid Tape