missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-SPP1 Matched Antibody Pair, Abnova
Brand: Abnova H00006696-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SPP1 (100μg); Mouse monoclonal anti-SPP1, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
ELISA
Specifications
SPP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SPP1. | |
BNSP/BSPI/ETA-1/MGC110940/OPN | |
Mouse, Rabbit | |
1 Set | |
Apoptosis | |
6696 | |
Human | |
IgG2a |
ELISA | |
Unconjugated | |
SPP1 | |
SPP1 | |
Affinity Purified | |
RUO | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SPP1 (100 μg) 2. Detection antibody: mouse monoclonal anti-SPP1, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |