missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNFSF11 Matched Antibody Pair, Abnova™
Brand: Abnova H00008600-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-TNFSF11, IgG1 Kappa (100μg); Detection antibody: biotinylated mouse monoclonal anti-TNFSF11, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF11.
Host Species: Mouse, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
TNFSF11 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF11. | |
CD254/ODF/OPGL/OPTB2/RANKL/TRANCE/hRANKL2/sOdf | |
Mouse | |
RUO | |
8600 | |
Human |
ELISA | |
Unconjugated | |
TNFSF11 | |
TNFSF11 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFSF11, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFSF11, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction