missing translation for 'onlineSavingsMsg'
Learn More

Antimony rod, 12.7mm (0.50in) dia x 10cm (3.9in) long, 99.8% (metals basis), Thermo Scientific Chemicals

1080.00 SEK - 3795.00 SEK

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485915
View Documents
Thermo Scientific Alfa Aesar
010098.KG
1 Pc.
1080.00 SEK
Each
Please to purchase this item. Need a web account? Register with us today!
15495915
View Documents
Thermo Scientific Alfa Aesar
010098.KJ
5 Pc.
3795.00 SEK
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]

Specifications

7440-36-0
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
99.8%
≈81.2g (0.178lbs)/10cm piece
(metals basis)
Sb
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
Odorless
Cast Rod
Antimony rod, 12.7mm (0.50 in.) dia. x 10cm (3.9 in.) long
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-146-5

missing translation for 'rtecsNumber' : CC4025000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.