missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AP180 Overexpression Lysate
Brand: Novus Biologicals™ NBP2-10304
6084.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The AP180 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_55656. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens AP180, mRNA. | |
Native |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction