missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Rabbit anti-Human, Mouse, Clone: 1A8N2, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Apolipoprotein L1 |
---|---|
Clone | 1A8N2 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321874
|
Bio-Techne
NBP3-16407-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18336914
|
Bio-Techne
NBP3-16407-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Apolipoprotein L1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8542 | |
IgG | |
Affinity purified |
1A8N2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 303-398 of human Apolipoprotein L1 (O14791). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title