Learn More
Apoptozole, MedChemExpress
MedChemExpress Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.
Brand: Medchem Express HY-15098
This item is not returnable.
View return policy
Description
Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.Specifications
Apoptozole | |
White | |
C33H25F6N3O3 | |
Apoptosis Activator VII | |
NC(C(C=C1)=CC=C1CN2C(C3=CC=C(OC)C=C3)=C(C4=CC=C(OC)C=C4)N=C2C5=CC(C(F)(F)F)=CC(C(F)(F)F)=C5)=O | |
625.56 | |
Research | |
Cancer-programmed cell death |
1054543-47-3 | |
Research | |
10 mg | |
DMSO : ≥ 100 mg/mL (159.86 mM) | |
625.56 | |
99.81% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.