Learn More
Apoptozole, MedChemExpress
MedChemExpress Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.
Brand:  Medchem Express HY-15098
Encompass_Preferred
 View more versions of this product
Product Code. 16426318
  • 1524.93 SEK / 10mg
Explore more special offers
Description
Description
Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.
Specifications
Specifications
Apoptozole
White
C33H25F6N3O3
Apoptosis Activator VII
NC(C(C=C1)=CC=C1CN2C(C3=CC=C(OC)C=C3)=C(C4=CC=C(OC)C=C4)N=C2C5=CC(C(F)(F)F)=CC(C(F)(F)F)=C5)=O
625.56
Research
Cancer-programmed cell death
1054543-47-3
Research
10 mg
DMSO : ≥ 100 mg/mL (159.86 mM)
625.56
99.81%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.