missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 2 Monoclonal Antibody (7H8B5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA531630
This item is not returnable.
View return policy
Description
Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.Specifications
Aquaporin 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P41181 | |
AQP2 | |
Purified recombit fragment of human AQP2 (AA: 149-271) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
7H8B5 | |
Unconjugated | |
AQP2 | |
AQP-CD; WCH-CD | |
Mouse | |
Protein G | |
RUO | |
359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |