missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arachidic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.
Solubility
Soluble in chloroform, ether, ethanol, and DMF. Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.
Solubility
Soluble in chloroform, ether, ethanol, and DMF. Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 74°C to 76°C |
Quantity | 1 g |
Merck Index | 14,764 |
Solubility Information | Soluble in chloroform,ether,ethanol,and DMF. Insoluble in water. |
Formula Weight | 312.54 |
Percent Purity | 98% |
Chemical Name or Material | Arachidic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'