missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Argon Gloves
Click to view available options
Size:
9
11
Description
- Suitable for argon welding and fine handling.
- Goat leather grain glove construction offers ideal mechanical resistance for welders.
- Sewn-on, one-piece thumb.
- Good flexibility and dexterity.
- Split leather cuff protects the forearm.
Specifications
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Attestation Number 0075/797/162/11/06/0313 |
Classification | Dexterity: 5 |
Cuff Material | Split leather |
Cut Resistance Rating | 1 |
Material | Grain Leather, Leather, Split Goat Leather |
Product Type | Welding Gloves |
Size | 11 |
Thickness (Metric) | 0.9 to 1 mm |
For Use With (Application) | Automotive, Building, Construction and Welding industries |
Puncture Rating | 1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'