missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ARMC8 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890484
|
Proteintech
12653-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880484
|
Proteintech
12653-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARMC8 is a protein coding gene.Specifications
ARMC8 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q8IUR7, Q9DBR3 | |
25852, 315949, 74125 | |
ARMC8 Fusion Protein Ag3374 | |
Primary | |
-20°C | |
ARMC8 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
armadillo repeat containing 8, ARMC8, HSPC056, S863 2 | |
Armc8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |