Learn More
WTW™ Armoring with Protective Hood for WTW™ ConOx™ and Tetracon™
The universal arming for the TetraCon™ 325 as well as for the galvanized oxygen sensor CellOx™ 325 provide optimal protection against damage in rough field applications, such as streaming water or bore holes.The special arming A pHLab/K is used to protect SenTix™ pH electrodes with glass shafts when measuring outside the lab.
3320.00 SEK - 3345.00 SEK
Specifications
For Use With (Equipment) | Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 |
---|---|
Type | Armoring |
Product Code | Brand | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
10613922
|
WTW™
903830 |
Plastic | Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 |
3320.00 SEK
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||||
10567831
|
WTW™
903831 |
Stainless Steel | Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- SensoLyt™ 900-P
- SensoLyt™ ORP 900-P
- TetraCon™ 925-P
- FDO™ 925-P
- VisoTurb™ 900-P
Specifications
Oxygen Sensor CellOx™ 325 and Conductivity Cell TetraCon™ 325 | |
Armoring |