Learn More
Arsenic AAS Standard, 1000 mg/L As in HCl 2%, Honeywell Fluka™
390.75 SEK - 691.09 SEK
Specifications
Chemical Name or Material | Arsenic AAS Standard |
---|---|
Density | 1.01 g/cm3 |
pH | <2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16624264
|
Honeywell Fluka
E4003-100ML |
100 mL |
390.75 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16634264
|
Honeywell Fluka
E4003-500ML |
500 mL |
691.09 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Arsenic AAS Standard | |
1.01 g/cm3 | |
<2 |
Safety and Handling
missing translation for 'shelfLife' : 760