missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSF Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | ARSF |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18322264
|
Bio-Techne
NBP3-04502-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18332874
|
Bio-Techne
NBP3-04502-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARSF Polyclonal specifically detects ARSF in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ARSF | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
416 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
arylsulfatase F, ASF, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ARSF (NP_004033.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title